The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026295.2 | NP_080571.2 | 12 - 13 | 2866 | 91 |
GenBank mRNA | AK016213.1 | - | 3 - 4 | 388 | 91 |
AK017665.1 | - | 1 - 2 | 75 | 91 | |
AK156617.1 | - | 12 - 13 | 2895 | 91 | |
BC053435.1 | - | 12 - 13 | 2866 | 91 |