Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex14
Entrez Gene ID
56273
Gene Name
peroxisomal biogenesis factor 14
Gene Aliases
Pex14p, R75137
Chromosome Location
Chr.4: 148960535 - 149099812 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019781.2 NP_062755.1 3 - 4 177 76
GenBank mRNA AF097512.1 - 3 - 4 177 76
AK148779.1 - 3 - 4 181 76
AK210926.1 - 1 - 2 49 77
BC028952.1 - 3 - 4 174 76

View Full Product Details