Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex14
Entrez Gene ID
56273
Gene Name
peroxisomal biogenesis factor 14
Gene Aliases
Pex14p, R75137
Chromosome Location
Chr.4: 148960535 - 149099812 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019781.2 NP_062755.1 8 - 9 688 124
GenBank mRNA AF097512.1 - 8 - 9 688 124
BC028952.1 - 8 - 9 685 124

View Full Product Details