Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
17756
Gene Name
microtubule-associated protein 2
Gene Aliases
G1-397-34, MAP-2, Mtap-2, Mtap2, repro4
Chromosome Location
Chr.1: 66175246 - 66442583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK141458.1 - 7 - 8 4409 124
M21041.1 - 7 - 8 124

View Full Product Details