The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008562.3 | NP_032588.1 | 2 - 3 | 1012 | 64 |
GenBank mRNA | AK010424.1 | - | 1 - 2 | 187 | 64 |
AK159504.1 | - | 2 - 3 | 924 | 64 | |
AK160594.1 | - | 1 - 2 | 184 | 64 | |
BC003839.1 | - | 2 - 3 | 941 | 64 | |
BC005427.1 | - | 2 - 3 | 934 | 64 | |
BC021638.1 | - | 2 - 3 | 951 | 64 | |
GU182318.1 | - | 3 - 4 | 738 | 64 | |
U35623.1 | - | 2 - 3 | 924 | 64 |