The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027864.1 | 7 - 8 | 1316 | 127 | |
GenBank mRNA | AK045495.1 | - | 7 - 8 | 1086 | 127 |
BC029728.1 | - | 7 - 8 | 1033 | 127 |