The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301705.1 | NP_001288634.1 | 3 - 4 | 375 | 57 |
NM_144899.3 | NP_659148.2 | 3 - 4 | 257 | 57 | |
GenBank mRNA | AK155481.1 | - | 3 - 4 | 305 | 57 |
AY158701.1 | - | 3 - 4 | 198 | 57 | |
BC117925.1 | - | 2 - 3 | 170 | 57 | |
BC117926.1 | - | 2 - 3 | 171 | 57 |