Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adamtsl4
Entrez Gene ID
229595
Gene Name
ADAMTS-like 4
Gene Aliases
ADAMTSL-4, Tsrc1
Chromosome Location
Chr.3: 95676201 - 95688444 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301705.1 NP_001288634.1 4 - 5 711 69
NM_144899.3 NP_659148.2 4 - 5 593 69
GenBank mRNA AK155481.1 - 4 - 5 641 69
AY158701.1 - 4 - 5 534 69
BC117925.1 - 3 - 4 506 69

View Full Product Details