Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
227059
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
2900042E17Rik, 5430433I10, mKIAA1265
Chromosome Location
Chr.1: 46807544 - 46925772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172653.2 NP_766241.2 5 - 6 1635 116
GenBank mRNA AK049099.1 - 3 - 4 381 116
AK122483.1 - 4 - 5 1069 116
AK136117.1 - 4 - 5 1347 116
BC052880.1 - 5 - 6 1749 116
BC059214.1 - 3 - 4 519 116
BC062918.1 - 5 - 6 1635 116

View Full Product Details