The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024264.4 | NP_077226.2 | 8 - 9 | 1540 | 61 |
GenBank mRNA | AK004977.1 | - | 8 - 9 | 1541 | 61 |
BC002183.1 | - | 7 - 8 | 1214 | 61 | |
BC055028.1 | - | 8 - 9 | 1518 | 61 |