The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008985.2 | NP_033011.2 | 22 - 23 | 2956 | 55 |
GenBank mRNA | AF332083.1 | - | 19 - 20 | 2560 | 55 |
AF332084.1 | - | 19 - 20 | 2551 | 55 | |
AK041296.1 | - | 22 - 23 | 2962 | 55 | |
AK076223.1 | - | 22 - 23 | 2937 | 55 | |
AK139398.1 | - | 21 - 22 | 3476 | 55 | |
AK220467.1 | - | 22 - 23 | 2914 | 55 | |
BC064020.1 | - | 10 - 11 | 1005 | 55 | |
BC068165.1 | - | 22 - 23 | 2888 | 55 | |
DQ832283.1 | - | 22 - 23 | 2933 | 55 | |
U11812.1 | - | 22 - 23 | 2874 | 55 | |
X74438.1 | - | 22 - 23 | 2973 | 55 |