Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf25
Entrez Gene ID
57751
Gene Name
ring finger protein 25
Gene Aliases
0610009H16Rik, AO7
Chromosome Location
Chr.1: 74593748 - 74601431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF171060.1 - 8 - 9 667 69
AK002399.1 - 8 - 9 688 69
AK011171.1 - 5 - 6 396 69
AK019236.1 - 5 - 6 391 69
AK171752.1 - 8 - 9 665 69
BC022715.1 - 8 - 9 653 69
BC037100.1 - 8 - 9 675 69
BC048821.1 - 8 - 9 675 69
BE853767.1 - 2 - 3 160 69

View Full Product Details