The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013913.3 | NP_038941.1 | 4 - 5 | 882 | 74 |
GenBank mRNA | AF162224.1 | - | 4 - 5 | 850 | 74 |
AK040679.1 | - | - | 277 | 74 | |
AK149450.1 | - | 4 - 5 | 883 | 74 | |
AK149555.1 | - | 4 - 5 | 883 | 74 | |
BC019491.1 | - | 4 - 5 | 870 | 74 |