Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf1
Entrez Gene ID
230584
Gene Name
Yip1 domain family, member 1
Gene Aliases
C030002N13Rik
Chromosome Location
Chr.4: 107314254 - 107359823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205156.1 NP_001192085.1 1 - 2 97 69
NM_145550.3 NP_663525.1 1 - 2 97 69
GenBank mRNA AK077502.1 - 1 - 2 74 69
AK081205.1 - 1 - 2 84 69
AK138108.1 - 1 - 2 92 69
AK157536.1 - 1 - 2 84 69
BY204210.1 - 1 - 2 70

View Full Product Details