The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165949.1 | NP_001159421.1 | 1 - 2 | 117 | 72 |
NM_018812.2 | NP_061282.2 | 1 - 2 | 117 | 72 | |
GenBank mRNA | BC050988.1 | - | 1 - 2 | 76 | 72 |