Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pias3
Entrez Gene ID
229615
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
Pias3l
Chromosome Location
Chr.3: 96696375 - 96706070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165949.1 NP_001159421.1 4 - 5 672 68
NM_018812.2 NP_061282.2 5 - 6 567 68
NM_146135.2 NP_666247.1 4 - 5 709 68
GenBank mRNA AF034080.3 - 4 - 5 444 68
AK040619.1 - 4 - 5 700 68
AK077551.1 - 4 - 5 658 68
BC023128.1 - 4 - 5 615 68
BC050988.1 - 5 - 6 526 68
BC051252.1 - 4 - 5 582 68

View Full Product Details