Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnlip
Entrez Gene ID
69060
Gene Name
pancreatic lipase
Gene Aliases
1810007A24Rik, PL, PTL
Chromosome Location
Chr.19: 58670365 - 58681788 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026925.3 NP_081201.2 7 - 8 719 54
GenBank mRNA AY387690.1 - 6 - 7 691 54
BC061061.1 - 7 - 8 719 54

View Full Product Details