Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx27
Entrez Gene ID
76742
Gene Name
sorting nexin family member 27
Gene Aliases
5730552M22Rik, ESTM45, ESTM47, R75405
Chromosome Location
Chr.3: 94497542 - 94582716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082484.2 NP_001075953.1 4 - 5 954 89
NM_029721.2 NP_083997.1 4 - 5 954 89
GenBank mRNA AK122296.1 - 4 - 5 766 89
AK147452.1 - 4 - 5 941 89
BC053495.1 - 4 - 5 766 89

View Full Product Details