The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172628.2 | NP_766216.2 | 2 - 3 | 198 | 70 |
GenBank mRNA | AK048066.1 | - | 2 - 3 | 199 | 70 |
AK052534.1 | - | 2 - 3 | 190 | 70 | |
AK143600.1 | - | - | 199 | 70 | |
AK178793.1 | - | 1 - 2 | 60 | 70 | |
AK194206.1 | - | 1 - 2 | 60 | 70 | |
AK194888.1 | - | 1 - 2 | 60 | 70 | |
AK198640.1 | - | 1 - 2 | 60 | 70 | |
BB841853.1 | - | 2 - 3 | 198 | 70 |