The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026470.3 | NP_080746.3 | 10 - 11 | 1293 | 91 |
GenBank mRNA | AK005819.1 | - | 5 - 6 | 680 | 91 |
AK006861.1 | - | 5 - 6 | 665 | 91 | |
AK051574.1 | - | 10 - 11 | 1294 | 91 | |
AK132771.1 | - | 10 - 11 | 1267 | 91 | |
AK153098.1 | - | 10 - 11 | 1170 | 91 | |
BC028429.1 | - | 5 - 6 | 693 | 91 | |
BC040024.1 | - | 5 - 6 | 693 | 91 |