Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Stradb
- Entrez Gene ID
-
227154
- Gene Name
- STE20-related kinase adaptor beta
- Gene Aliases
-
AA792893, Als2cr2, B830008M19, D1Ucla2, ILPIP, ILPIPA, PRO1038, Papk, Syradb
- Chromosome Location
-
Chr.1: 58973527 - 58995126 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB057667.1 |
- |
7 - 8 |
680 |
94 |
AK140272.1 |
- |
7 - 8 |
963 |
94 |
View Full Product Details