Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ndst1
- Entrez Gene ID
-
15531
- Gene Name
- N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1
- Gene Aliases
-
1200015G06Rik, Hsst, NDST-1, b2b2230Clo
- Chromosome Location
-
Chr.18: 60684494 - 60748892 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK170041.1 |
- |
1 - 2 |
85 |
59 |
AK171013.1 |
- |
1 - 2 |
85 |
59 |
View Full Product Details