The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026854.3 | NP_081130.1 | 4 - 5 | 658 | 112 |
GenBank mRNA | AK004450.1 | - | 4 - 5 | 633 | 112 |
AK144689.1 | - | 4 - 5 | 633 | 112 | |
BC075693.1 | - | 6 - 7 | 752 | 112 | |
BC139347.1 | - | 4 - 5 | 624 | 112 | |
BC139348.1 | - | 4 - 5 | 624 | 112 |