The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113559.2 | NP_001107031.2 | 1 - 2 | 237 | 87 |
NM_001243163.1 | NP_001230092.1 | 1 - 2 | 237 | 87 | |
NM_011444.3 | NP_035574.2 | 1 - 2 | 237 | 87 | |
GenBank mRNA | AB006330.1 | - | 1 - 2 | 149 | 87 |
AJ010604.1 | - | 1 - 2 | 227 | 87 | |
AK132307.1 | - | 1 - 2 | 130 | 87 | |
BB655610.1 | - | 1 - 2 | 238 | 87 | |
BC110478.2 | - | 1 - 2 | 121 | 87 | |
KF793916.1 | - | 1 - 2 | 43 | 87 |