Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhg5
Entrez Gene ID
269608
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 5
Gene Aliases
BC023181, Syx
Chromosome Location
Chr.4: 152072433 - 152115404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285999.1 NP_001272928.1 20 - 21 3280 122
NR_104381.1 21 - 22 3246 122
GenBank mRNA AK129198.1 - 7 - 8 1488 122
AK138147.1 - 6 - 7 2229 122
AK154912.1 - 21 - 22 3239 122
AY605057.1 - 20 - 21 3280 122
BC023181.1 - 6 - 7 1426 122
BC064091.1 - 2 - 3 881 122

View Full Product Details