Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agrn
Entrez Gene ID
11603
Gene Name
agrin
Gene Aliases
Agrin, nmf380
Chromosome Location
Chr.4: 156165290 - 156197519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021604.3 NP_067617.3 29 - 30 5172 74
GenBank mRNA AK147577.1 - 27 - 28 5605 74
AY914875.1 - 11 - 12 1896 74
BC028992.1 - 1 - 2 53 74
BC059259.1 - 16 - 17 2586 74
BC150703.1 - 27 - 28 4626 74

View Full Product Details