Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agrn
Entrez Gene ID
11603
Gene Name
agrin
Gene Aliases
Agrin, nmf380
Chromosome Location
Chr.4: 156165290 - 156197519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021604.3 NP_067617.3 1 - 2 276 97
GenBank mRNA AF190858.1 - 1 - 2 276 97
AK134826.1 - 1 - 2 289 97
AY914875.1 - 1 - 2 219 97
U84407.1 - 1 - 2 130 97

View Full Product Details