Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhn1
Entrez Gene ID
231002
Gene Name
pleckstrin homology domain containing, family N member 1
Gene Aliases
BC025458
Chromosome Location
Chr.4: 156221456 - 156228542 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008233.3 NP_001008234.2 14 - 15 1734 61
GenBank mRNA AK048084.1 - 12 - 13 3621 61
BC025458.1 - 4 - 5 605 61
BC046544.1 - - 1896 61

View Full Product Details