The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008233.3 | NP_001008234.2 | 15 - 16 | 1780 | 80 |
GenBank mRNA | AK048084.1 | - | 13 - 14 | 3667 | 80 |
BC025458.1 | - | 5 - 6 | 651 | 80 |