The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008233.3 | NP_001008234.2 | 8 - 9 | 856 | 99 |
GenBank mRNA | AK048084.1 | - | 6 - 7 | 2835 | 99 |
BC046544.1 | - | 8 - 9 | 856 | 99 |