Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npc1
Entrez Gene ID
18145
Gene Name
Niemann-Pick type C1
Gene Aliases
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm
Chromosome Location
Chr.18: 12189694 - 12236386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008720.2 NP_032746.2 7 - 8 1106 66
GenBank mRNA AF003348.1 - 7 - 8 1083 66
AK154282.1 - 7 - 8 1026 66
AK155379.1 - 7 - 8 1026 66
BC052437.1 - 7 - 8 1106 66
BC054539.1 - 7 - 8 1106 66

View Full Product Details