The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190371.1 | NP_001177300.1 | 7 - 8 | 715 | 89 |
GenBank mRNA | AK162301.1 | - | 2 - 3 | 143 | 89 |