Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrfn5
Entrez Gene ID
238205
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
AI427653, AI604817, C130061B21, mKIAA4208
Chromosome Location
Chr.12: 61522817 - 61857930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178714.4 NP_848829.2 2 - 3 376 106
GenBank mRNA AK034245.1 - 2 - 3 377 106
AK048443.1 - 2 - 3 462 106
AK083920.1 - 2 - 3 1570 107
AK220546.1 - 2 - 3 1060 106
BC052038.1 - 2 - 3 206 106

View Full Product Details