Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dus4l
Entrez Gene ID
71916
Gene Name
dihydrouridine synthase 4-like (S. cerevisiae)
Gene Aliases
2310069P03Rik, 2700089B10Rik, AI482040, Pp35
Chromosome Location
Chr.12: 31640050 - 31654826 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028002.2 NP_082278.1 2 - 3 283 142
GenBank mRNA AK010138.1 - 2 - 3 284 142
BC109351.2 - 2 - 3 218 142

View Full Product Details