Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp5
Entrez Gene ID
240672
Gene Name
dual specificity phosphatase 5
Gene Aliases
Gm337
Chromosome Location
Chr.19: 53529318 - 53541322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085390.1 NP_001078859.1 1 - 2 378 69

View Full Product Details