The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008072.2 | NP_032098.2 | 8 - 9 | 1167 | 77 |
GenBank mRNA | AF326546.1 | - | 8 - 9 | 1060 | 77 |
AF326564.1 | - | 8 - 9 | 1060 | 77 | |
BC116832.1 | - | 8 - 9 | 1102 | 77 | |
BC116836.1 | - | 8 - 9 | 1102 | 77 | |
S42882.1 | - | 8 - 9 | 1160 | 77 |