Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl2
Entrez Gene ID
52686
Gene Name
methyltransferase like 2
Gene Aliases
2810438F06Rik, AW493404, C130031G21Rik, D11Ertd768e, METL, PSENIP1
Chromosome Location
Chr.11: 105126425 - 105141146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172567.3 NP_766155.3 6 - 7 864 98
GenBank mRNA AK030739.1 - 5 - 6 1554 98
AK048040.1 - 6 - 7 865 98
AK133887.1 - 5 - 6 1554 98
AK153431.1 - 6 - 7 798 98
BC089591.1 - 6 - 7 802 98

View Full Product Details