The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271716.1 | NP_001258645.1 | 1 - 2 | 108 | 137 |
NM_001301372.1 | NP_001288301.1 | 1 - 2 | 108 | 137 | |
GenBank mRNA | D85688.1 | - | 1 - 2 | 108 | 137 |