The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008791.2 | NP_001008791.1 | 12 - 13 | 3334 | 59 |
NM_001008792.2 | NP_001008792.1 | 11 - 12 | 3313 | 59 | |
NM_001008793.2 | NP_001008793.1 | 11 - 12 | 3298 | 59 | |
NM_028640.2 | NP_082916.1 | 11 - 12 | 3301 | 59 | |
GenBank mRNA | AK122523.1 | - | 11 - 12 | 2429 | 59 |
AY739114.1 | - | 11 - 12 | 3261 | 59 | |
AY739115.1 | - | 12 - 13 | 3294 | 59 | |
AY739116.1 | - | 11 - 12 | 3273 | 59 | |
AY739117.1 | - | 11 - 12 | 3258 | 59 | |
AY739118.1 | - | 8 - 9 | 1910 | 59 | |
AY739119.1 | - | 7 - 8 | 1877 | 59 | |
AY739120.1 | - | 6 - 7 | 1664 | 59 | |
AY739121.1 | - | 6 - 7 | 1782 | 59 | |
AY739122.1 | - | 7 - 8 | 2114 | 59 | |
BC150757.1 | - | 11 - 12 | 2658 | 59 |