Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem61
Entrez Gene ID
329909
Gene Name
transmembrane protein 61
Gene Aliases
4732441L20Rik
Chromosome Location
Chr.4: 106498813 - 106506335 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK028711.1 - 1 - 2 693 83

View Full Product Details