Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d5
Entrez Gene ID
72238
Gene Name
TBC1 domain family, member 5
Gene Aliases
1600014N05Rik
Chromosome Location
Chr.17: 50733124 - 51180198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285991.1 NP_001272920.1 6 - 7 714 127
NM_001285993.1 NP_001272922.1 6 - 7 681 127
NM_028162.4 NP_082438.3 6 - 7 714 127
GenBank mRNA AK005461.1 - 6 - 7 698 127
AK033710.1 - 6 - 7 606 127
AK144467.1 - 6 - 7 715 127
AK154975.1 - 6 - 7 674 127
AK163366.1 - 5 - 6 523 127
AK220251.1 - 6 - 7 681 127
BC043113.1 - 6 - 7 530 127
BC098328.1 - 6 - 7 546 127

View Full Product Details