The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205369.1 | NP_001192298.1 | 9 - 10 | 1470 | 74 |
NM_001205371.1 | NP_001192300.1 | 9 - 10 | 1060 | 74 | |
NM_177054.5 | NP_796028.2 | 8 - 9 | 1371 | 74 | |
GenBank mRNA | AK131751.1 | - | 9 - 10 | 1060 | 74 |
AY484583.1 | - | 8 - 9 | 1211 | 74 | |
BC064446.1 | - | 9 - 10 | 1470 | 74 |