Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec24c
Entrez Gene ID
218811
Gene Name
Sec24 related gene family, member C (S. cerevisiae)
Gene Aliases
2610204K03Rik, mKIAA0079
Chromosome Location
Chr.14: 20674285 - 20694852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168273.1 NP_001161745.1 13 - 14 1671 102
NM_172596.2 NP_766184.2 12 - 13 1899 102
GenBank mRNA AK036519.1 - 12 - 13 1899 102
AK122199.1 - 13 - 14 1649 102
AK132090.1 - 7 - 8 926 102
BC027775.1 - 2 - 3 274 102
BC040370.1 - 12 - 13 1877 102

View Full Product Details