Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd16
Entrez Gene ID
330660
Gene Name
BTB (POZ) domain containing 16
Gene Aliases
E330040A16Rik
Chromosome Location
Chr.7: 130774069 - 130825899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081038.2 NP_001074507.2 9 - 10 1184 68

View Full Product Details