The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163703.1 | NP_001157175.1 | 2 - 3 | 722 | 76 |
NM_173408.3 | NP_775584.1 | 2 - 3 | 741 | 76 | |
GenBank mRNA | AK029043.1 | - | 2 - 3 | 687 | 76 |
AK155160.1 | - | 2 - 3 | 723 | 76 | |
BC030335.1 | - | 2 - 3 | 664 | 76 |