Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgr4
Entrez Gene ID
107515
Gene Name
leucine-rich repeat-containing G protein-coupled receptor 4
Gene Aliases
9130225G07, A330106J01Rik, A930009A08Rik, Gpr48
Chromosome Location
Chr.2: 109917647 - 110014257 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172671.2 NP_766259.2 14 - 15 1732 77
GenBank mRNA AK044357.1 - 13 - 14 1040 77
CN667965.1 - 5 - 6 304 77

View Full Product Details