Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcte1
Entrez Gene ID
21645
Gene Name
t-complex-associated testis expressed 1
Gene Aliases
D17Sil1, Tcte-1
Chromosome Location
Chr.17: 45523434 - 45542679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013688.2 NP_038716.2 2 - 3 505 73
GenBank mRNA BC145741.1 - 1 - 2 312 73
CX237405.1 - 2 - 3 398 73
M28821.1 - 1 - 2 312 73

View Full Product Details