Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fnbp4
Entrez Gene ID
55935
Gene Name
formin binding protein 4
Gene Aliases
FBP30, Fnbp30, mKIAA1014
Chromosome Location
Chr.2: 90745370 - 90781021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018828.2 NP_061298.1 3 - 4 691 56
GenBank mRNA AK012167.1 - 3 - 4 504 56
AK014018.1 - 3 - 4 508 56
AK084018.1 - 3 - 4 483 56
AK129263.1 - 3 - 4 588 56
AK164543.1 - 3 - 4 462 56
U40750.2 - 3 - 4 691 56

View Full Product Details