Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pla2g4b
- Entrez Gene ID
-
211429
- Gene Name
- phospholipase A2, group IVB (cytosolic)
- Gene Aliases
-
A030011C02Rik, Gm1013, cPLA2-beta
- Chromosome Location
-
Chr.2: 120033433 - 120043033 on Build GRCm38
- Gene Symbol
- Gm28042
- Entrez Gene ID
-
102238433
- Gene Name
- predicted gene, 28042
- Gene Aliases
-
Jmjd7-pla2g4b
- Chromosome Location
-
Chr.2: 120027483 - 120043033 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104353.1 |
|
18 - 19 |
2215 |
69 |
GenBank mRNA |
AB195274.1 |
- |
18 - 19 |
2205 |
69 |
AB195275.1 |
- |
17 - 18 |
4313 |
69 |
BC098210.1 |
- |
12 - 13 |
1366 |
69 |
View Full Product Details