Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Stard9
- Entrez Gene ID
-
668880
- Gene Name
- START domain containing 9
- Gene Aliases
-
4831403C07Rik, E230025N21Rik, Kif16a, mKIAA1300
- Chromosome Location
-
Chr.2: 120629091 - 120716086 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK087617.1 |
- |
1 - 2 |
452 |
83 |
View Full Product Details